missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ QCM Chemotaxis Cell Migration Assay, 24-well (5μm), fluorimetric

Activity Assay

Supplier:  MilliporeSigma™ ECM507

Catalog No. ECM507


Only null left

Description

Description

Specifications

Specifications

2°C to 8°C
Fluorescence
Forceps, Sterile 3μm 24-well Cell migration plate, two 24-well plates w/12 inserts/plate, Cell detachment soln., 4X cell lysis buffer, CyQUANT GR Dye
QCM Chemotaxis Cell Migration Assay
Activity Assay
1/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions