missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ QCM 3um Endothelial Cell Migration Assay Fibronectin, Fluorimetric

Catalog No. ECM201
Change view
Click to view available options
Quantity:
1/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ECM201 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ECM201 Supplier MilliporeSigma™ Supplier No. ECM201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For the study of endothelial cell migration in response to an angiogenic stimulus

Specifications

Content And Storage -20°C
Description Endothelial Cell Migration Assay Fibronectin
Detection Method Fluorescence
For Use With (Application) For the study of endothelial cell migration in response to an angiogenic stimulus
Includes Accutase, Fibronectin test plate, BSA control plate, 4X Cell Lysis Buffer, CyQuant GR Dye; Forceps, 24-well/96-Well plates
Quantity 1/Pk.
Type Cell-Based Assay Kit
Druggable Target Cell-based Assay