missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ PARP1 Enzyme Activity Assay

Catalog No. 1710149
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1710149 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1710149 Supplier MilliporeSigma™ Supplier No. 1710149

For quantifying PARP1 and PARP2 activity

Specifications

Content And Storage Maintain the unopened kit at -80°C
Description PARP1 Enzyme Activity Assay
Detection Method Fluorescence
For Use With (Application) For quantifying PARP1 and PARP2 activity
Includes Black 96 well plate - 1 plate, PARP1 Assay Buffer - 4mL, Plate Covers - 2 plate covers, Activated DNA - 1 vial (100μg at 10mg/mL), β-NAD - 1 vial (30μL at 50mM), Nicotinamide (NAM) - 1 vial (10μL at 10mM), Recombinant PARP1 Enzyme - 1 vial (2μg at 0.2μg/μL), Recombinant Nicotinamidase Enzyme with 50% glycerol - 1 vial (60μg at 1mg/mL), 3-ABA (PARP Inhibitor) in DMSO - 1 vial (300μL at 100mM), Developer Reagent - 4mL at 10mM.
Quantity 96 Assays