missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ OxyICC™ Oxidized Protein Detection Kit

Catalog No. S7350
Change view
Click to view available options
Quantity:
40 Reactions in an 8 Chamber Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S7350 40 Reactions in an 8 Chamber Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S7350 Supplier MilliporeSigma™ Supplier No. S7350

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For immunocytochemistry, immunofluorescence

Specifications

Content And Storage Dual temperature storage. Store for up to 6 months after date of receipt.
Description OxyICC Oxidized Protein Detection Kit
Detection Method Fluorescence
For Use With (Application) Immunocytochemistry, Immunofluorescence
Includes Streptavidin-Cy3, 2,4-Dinitrophenylhydrazine Solution, 10x PBS wash buffer, 5x blocking buffer, Anti-DNP, Biotinylated (mouse monoclonal) antibody, DAPI
Quantity 40 Reactions in an 8 Chamber Slide