missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Neural Stem Cell Basal Medium

Catalog No. SCM003
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM003 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCM003 Supplier MilliporeSigma™ Supplier No. SCM003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

a defined serum-free, growth factor-free medium that has been optimized for the growth and in vitro differentiation of neural stem cells derived from rodents.

The Neural Stem Cell Basal Medium is a defined serum-free, growth factor-free medium that has been optimized for the growth and in vitro differentiation of neural stem cells derived from rodents. When used in conjunction with bFGF or bFGF, EGF and heparin, the basal medium will allow for the proliferation of rat and mouse neural stem cells, respectively. Withdrawal of the growth factors from the Neural Stem Cell Basal Medium will result in the spontaneous differentiation of rodent neural stem cells.

Specifications

For Use With (Application) Stem Cell Culture
Quantity 500 mL
Content And Storage −20°C