missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ MTT Reagent A

Catalog No. CT015
Change view
Click to view available options
Quantity:
5 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CT015 5 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CT015 Supplier MilliporeSigma™ Supplier No. CT015

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For study of cell and mitochondrial health

Specifications

Content And Storage 2°C to 8°C
Description MTT Reagent A
For Use With (Application) For study of cell and mitochondrial health
Quantity 5 Vials