missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Mast Cell Degranulation Assay Kit

Catalog No. IMM001
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IMM001 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IMM001 Supplier MilliporeSigma™ Supplier No. IMM001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Activity assay, Evaluation of tryptase activity in culture supernatants, cell lysates or other tryptase-containing samples

Specifications

Content And Storage Store kit materials (as provided) at -20°C up to their expiration date.
Description Mast cell degranulation assay kit
For Use With (Application) Activity assay, Evaluation of tryptase activity in culture supernatants, cell lysates or other tryptase-containing samples
Includes pNA standard, Tryptase positive control, 5X assay buffer , calcium ionophore A23187, Tryptase inhibitor (Protamine), Tryptase substrate
Quantity 96 Assays