missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Chemicon™ LentiBrite™ GFP-LC3 Lentiviral Biosensor
Description
- Supports monitoring of autophagosome formation by fluorescence microscopy.
Specifications
Specifications
| Content And Storage | Lentivirus is stable for at least 4 months from date of receipt when stored at -80°C. |
| Description | GFP-LC3 Lentiviral Biosensor |
| Detection Method | Fluorescence |
| For Use With (Application) | Immunocytochemistry, Transfection, Immunofluorescence |
| Includes | TagGFP2-LC3 Lentivirus: One vial containing 25μL of lentiviral particles at a minimum of 3 x 10E8 infectious units (IFU) per mL; Promoter: EF-1 (Elongation Factor-1) |
| Quantity | 1 Vial (minimum of 3 x 108 IFU/mL) |