missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ FibroGRO™ Xeno-Free Human Fibroblast Expansion Medium

Catalog No. SCM044
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM044 500 mL
SCM037 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SCM044 Supplier MilliporeSigma™ Supplier No. SCM044

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

a new variation of our FibroGRO cell culture media that is optimized for the culture of fibroblasts without the use of animal-derived components.

FibroGRO™ Xeno-Free Human Fibroblast Expansion Medium is a new variation of our FibroGRO™ cell culture media that is optimized for the culture of fibroblasts without the use of animal-derived components. EMD Millipore’s FibroGRO™ cell system is perfect for establishing Xeno-Free human feeder layers for induced pluripotent, embryonic, and general stem cell culture. It can also be used to establish a model to study wound healing, toxicology and basic cell biology. FibroGRO™ Xeno Free supports the growth of these cells in a 2% human serum environment at rates equal to or greater than comparable media supplemented with 2-10% FBS. Our FibroGRO™ Xeno-Free media does not contain any antimicrobials, phenol red, or any other components that can cause cell stress and “masking effects” that may influence experimental results.

Specifications

For Use With (Application) Cell Culture
Quantity 500 mL
Content And Storage -20°C