missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ FibroGRO™-LS Complete Media Kit

Catalog No. SCMF002
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCMF002 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCMF002 Supplier MilliporeSigma™ Supplier No. SCMF002

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

an optimal cell culture environment for many types of human fibroblasts and mouse embryonic fibroblasts.

FibroGRO™ Media formulations provide an optimal cell culture environment for many types of human fibroblasts and mouse embryonic fibroblasts These low-serum/serum-free formulations have been shown to grow these cells at rates that meet or exceed commercially available serum containing media, while maintaining excellent cell morphology. The media are packaged in a specially designed UV protective shrinkwrap for added stability, which includes a temperature gauge for added convenience. FibroGRO Media contain no antimicrobials and no phenol red, components that can cause cell stress and masking effects that may influence experimental results.

Specifications

For Use With (Application) Cell Culture
Quantity 500 mL
Content And Storage Basal medium: Store at 4°C in dark; Supplement Mix: -20