missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ EpiGRO™ Human Ocular Epithelia Complete Media Kit

Catalog No. SCMC001
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCMC001 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCMC001 Supplier MilliporeSigma™ Supplier No. SCMC001

a serum-free medium optimized for the culture of human corneal epithelial cells.

EpiGRO™ Human Ocular Epithelia Complete Media Kit is a serum-free medium optimized for the culture of human corneal epithelial cells. It supports the growth of these cells over a period of at least 3 passages. This medium does not contain any antimicrobials or phenol red; components that can cause cell stress and “masking effects” that may influence experimental results. It is formulated for use in an incubator with an atmosphere of 5% CO⒉

Specifications

For Use With (Application) Cell culture
Quantity 500 mL
Content And Storage Basal medium: 2-8°C in dark; Supplement Mix: -20