missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ EndoGRO-MV-VEGF Complete Culture Media Kit

Catalog No. SCME003
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCME003 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCME003 Supplier MilliporeSigma™ Supplier No. SCME003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

a low serum formulation, intended for rapid proliferation of human microvascular endothelial cells.

EndoGRO Media Formulations provide an optimal cell culture environment for many types of endothelial cells, including HUVEC, aortic endothelial cells and other human large vessel and microvascular endothelial cells. These media have been shown to grow endothelial cells at rates that meet or exceed commercially available serum containing media, while maintaining excellent cell morphology. The media are packaged in a specially designed UV protective shrinkwrap for added stability, which includes a temperature gauge that helps prevent contamination. EndoGRO Media contain no antimicrobials and no phenol red, components that can cause cell stress and masking effects that may influence experimental results. Four different media formulations are available for various applications and cell culture requirements.

Specifications

For Use With (Application) Cell culture
Quantity 500 mL
Content And Storage Basal medium: 2-8°C in dark; Supplement Mix: -20