missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Differentiation Kit

Catalog No. SCR600
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR600 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR600 Supplier MilliporeSigma™ Supplier No. SCR600

contains Human Oligodendrocyte Progenitor Cells (OPCs) along with optimized media for their expansion and spontaneous differentiation into mature oligodendrocytes.

EMD Millipore’s Human Oligodendrocyte Differentiation Kit contains Human Oligodendrocyte Progenitor Cells (OPCs) along with optimized media for their expansion and spontaneous differentiation into mature oligodendrocytes. Human Oligodendrocyte Progenitor Cells (OPCs) are derived from NIH approved H9 human embryonic stem cells (hESCs). These hESC-derived OPCs proliferate as an adherent cell monolayer and greater than 70% express the appropriate early-to-intermediate oligodendrocyte markers, including O4, NG2, Olig2, Sox10 and GalC. Upon growth factor withdrawal, Human OPCs spontaneously differentiate into mature oligodendrocytes expressing terminally differentiated markers including proteolipid protein (PLP), myelin basic protein (MBP), and myelin oligodendrocyte glycoprotein (MOG). Spontaneous differentiation leads to approximately 30% of the cells expressing PLP, MBP and MOG while the remaining 70% express mostly neuronal markers, MAP2 and beta III-tubulin with very little to no astrocytes (<5% GFAP-positive cells) observed. Human OPCs can be used for a variety of research applications including studies of neurotoxicity, co-culture applications and screening for molecules that induce or inhibit preferential differentiation to mature oligodendrocytes.

Specifications

For Use With (Application) Stem Cell Culture
Quantity 1 Kit
pH 7.4±0.4
Content And Storage Human Oligodendrocyte Progenitors (OPCs), 500,000 viable cells: (Mfr.. No. CS204496); Human OPC Spontaneous Differentiation Media Kit, 100mL: (Mfr.. No. SCM106); Human OPC Expansion Media Kit, 100mL: (Mfr. No. SCM107)