missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ CpGenome™ Universal Methylated DNA

Catalog No. S7821
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S7821 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S7821 Supplier MilliporeSigma™ Supplier No. S7821

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gene methylation studies

Specifications

Content And Storage -15°C to -25°C
Description CpGenome universal methylated DNA
For Use With (Application) Gene methylation studies
Quantity 10 μg