missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Chemicon™ CpG WIZ™ RARβ2-Methylation Specific PCR Assay
Specifications
Specifications
| Content And Storage | -15°C to -25°C |
| Description | CpG WIZ RARbeta2-methylation specific PCR assay |
| For Use With (Application) | PCR |
| Includes | Universal 10X PCR buffer, U primer set 7.5μm, M primer set 7.5μm, W primer set 7.5μm, U control DNA, M control DNA, W control DNA |
| Quantity | 25 Assays |
| Type | DNA Assay Kit |