Learn More
MilliporeSigma™ Chemicon™ CpG WIZ™ MGMT-Methylation Specific PCR Assay
Description
- Includes primer sets and control DNA for methylation-specific PCR.
Specifications
Specifications
| Description | CpG WIZ MGMT-methylation specific PCR assay |
| For Use With (Application) | PCR |
| Includes | Universal 10X PCR buffer, U primer set 5μm, M primer set 5μm, W primer set 5μm, U control DNA, M control DNA, W control DNA |
| Quantity | 25 Assays |