missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ CpG WIZ™ Fragile X Amplification Kit

Catalog No. S7807
Change view
Click to view available options
Quantity:
25 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S7807 25 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S7807 Supplier MilliporeSigma™ Supplier No. S7807

missing translation for 'validMainframeMsg'

For PCR amplification after bisulfite modification of DNA samples

  • Includes primer sets, control DNA and PCR buffer.

Specifications

Content And Storage -15°C to -25°C
Description CpG WIZ Fragile X amplification kit
For Use With (Application) PCR
Includes Universal 10X PCR buffer, U primer set 5μm, M primer set 5μm, W primer set 5μm, U control DNA, M control DNA, W control DNA
Quantity 25 Assays