missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ CpG WIZ™ ERα-methylation specific PCR assay

Catalog No. S7815
Change view
Click to view available options
Quantity:
25 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S7815 25 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S7815 Supplier MilliporeSigma™ Supplier No. S7815

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For PCR amplification after bisulfite modification of DNA samples

Specifications

Content And Storage -15°C to -25°C
Description CpG WIZ ERalpha-methylation specific PCR assay
For Use With (Application) PCR
Includes Universal 10X PCR buffer, U primer set 7.5μm, M primer set 7.5μm, W primer set 7.5μm, U control DNA, M control DNA, W control DNA
Quantity 25 Assays