missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ CpG WIZ™ BRCA1-Methylation Specific PCR Assay

Catalog No. S7830
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S7830 25 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. S7830 missing translation for 'mfr' MilliporeSigma™ missing translation for 'supplierNo' S7830

For PCR, sensitive detection of altered DNA

Specifications

Content And Storage -15°C to -25°C
Description CpG WIZ BRCA1-methylation specific PCR assay
For Use With (Application) PCR
Includes Universal 10X PCR buffer, U primer set 7.5μm, M primer set 7.5μm, W primer set 7.5μm, U control DNA, M control DNA, W control DNA
Quantity 25 Assays