missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Cellular senescence assay

Catalog No. KAA002
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KAA002 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KAA002 Supplier MilliporeSigma™ Supplier No. KAA002

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Efficiently detects SA-beta-gal activity at pH 6.0

Specifications

Content And Storage Store X-gal solution protected from light at -20°C, and other kit components at 4deg.C
Description Cellular senescence assay
Detection Method Chromogenic
For Use With (Application) Efficiently detects SA-beta-gal activity at pH 6.0
Includes X-gal solution, 100X fixing solution, 10X Staining Solution A, 10X Staining Solution B
Quantity 1 Kit