missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Chemicon™ beta-Integrin-mediated Cell Adhesion Array Kit, Fluorimetric
Specifications
Specifications
| Description | beta-Integrin-mediated Cell Adhesion Array Kit |
| Detection Method | Fluorescence |
| For Use With (Application) | For assessing the presence or absence of specific integrins on the cell surface |
| Includes | Assay Buffer, beta Integrin Array Plate, 4X-cell lysis buffer, CyQuant GR Dye |
| Quantity | 1/Pk. |
| Type | Cell-Based Assay Kit |
| Cell Line | Any Adherent Cell Type |
| Druggable Target | Cell-based Assay |