missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Chemicon™ ApopTag™ Plus In Situ Apoptosis Fluorescein Detection Kit
Specifications
Specifications
| Content And Storage | Store the kit at -15 to -25°C until the first use. After the first use, if the kit will be used within three months, store the TdT Enzyme at -15 to -25°C and store remaining components at 2-8°C |
| Description | ApopTag plus in situ apoptosis fluorescein detection kit |
| Detection Method | Fluorescence |
| For Use With (Application) | Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Includes | Affinity purified sheep polyclonal antibody, equilibration buffer, reaction buffer, TdT enzyme, stop/wash buffer, Anti-Digoxigenin-Fluorescein, blocking solution, plastic coverslips, control slides |
| Quantity | 40 Assays |