missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Apo-Direct TUNEL Assay Kit

Catalog No. APT110
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APT110 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APT110 Supplier MilliporeSigma™ Supplier No. APT110

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For labeling DNA breaks to detect apoptotic cells by flow cytometry

Specifications

Description Apo-Direct TUNEL Assay Kit
Detection Method Fluorescence
For Use With (Application) For labeling DNA breaks to detect apoptotic cells by flow cytometry
Includes PI/RNase Staining Buffer, Positive Control Cells, Negative Control Cells, Wash Buffer, Reaction Buffer, TdT Enzyme, F-dUTP, Rinsing Buffer
Quantity 50 Tests