missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Amplifluor™ SNPs HT Genotyping System FAM-SR

Catalog No. S7908
Change view
Click to view available options
Quantity:
5000 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S7908 5000 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S7908 Supplier MilliporeSigma™ Supplier No. S7908

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fluorescent plate readers

Specifications

Description Amplifluor SNPs HT genotyping system FAM-SR
Detection Method Fluorescence
For Use With (Application) Fluorescent plate readers
Quantity 5000 Assays