missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Amplifluor™ SNPs HT Genotyping System FAM-JOE

Catalog No. S7909
Change view
Click to view available options
Quantity:
5000 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S7909 5000 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S7909 Supplier MilliporeSigma™ Supplier No. S7909

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Real-time PCR detection instruments

Specifications

Description  Amplifluor SNPs HT genotyping system FAM-JOE
Detection Method Fluorescence
For Use With (Application) Real-time PCR detection instruments
Quantity 5000 Assays