missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Chemicon™ 3D Collagen Culture Kit
Description
- Provides a three-dimensional collagen matrix for cell culture.
- Cells can be visualized using phase-contrast or fluorescence microscopy.
- Supports fixation, staining and reagent treatment within the matrix.
Specifications
Specifications
| Content And Storage | Store kit materials at 4deg.C up to their expiration date |
| Description | 3D collagen culture kit |
| For Use With (Application) | Enzyme assay |
| Includes | Neutralization/Collagen Solution, 5X RPMI/M199/DMEM Medium, PBS with Phenol Red |
| Quantity | 1 Kit |
| Type | Reagent Kit |