missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ 20S Proteasome Activity Assay Kit

Catalog No. APT280
Change view
Click to view available options
Quantity:
1/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APT280 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APT280 Supplier MilliporeSigma™ Supplier No. APT280

Provides a simple and convenient means for assaying proteasome activity that recognize the substrate LLVY

Specifications

Content And Storage -20°C
Description 20S Proteasome Activity Assay Kit
For Use With (Application) Provides a simple and convenient means for assaying proteasome activity that recognize the substrate LLVY
Includes AMC Standard, 20S Proteasome Positive Control, 10X Assay Buffer, Proteasome Substrate, Lactacystin
Quantity 1/Pk.