missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Certistain™ Hemotoxylin Solution

Catalog No. M1051742500
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1051742500 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1051742500 Supplier MilliporeSigma™ Supplier No. 1051742500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

for microscopy

In hematoxylin-eosin staining (HandE), the most widely used histological staining technique, nuclear staining is achieved using ready-to-use staining solutions, such as Mayer’s hemalum solution or Hematoxylin solution modified according to Gill III. These solutions do not need to be filtrated prior use and the dye is oxidized in a controlled manner, which renders the results robust and allows a brilliant differentiation of the stained structures. Counterstaining of i.e. for proteins, collagen, keratin, intercellular substances. can be performed with alcoholic or aqueous Eosin Y solution 0.5 %. A 500mL bottle is sufficient for 1500 up to 2500 slides. It is registered as IVD and CE product.

Specifications

Color Reddish-violet
Grade Analytical
Product Type Hematoxylin
Quantity 2.5 L
Solution Type Stain
Form Liquid