missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Catalase, Aspergillus niger, Protein

Catalog No. 219261100KU
Change view
Click to view available options
:
100kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
219261100KU 100kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 219261100KU Supplier MilliporeSigma™ Supplier No. 219261100KU
Only null left
Native catalase from Aspergillus niger. Long-acting, extremely stable form of catalase that is active over a wide pH range (pH 1 - 12). Composed of four protein subunits, each containing a heme group bound to its active site.

Specifications

Preparation Method H2O (1mg/mL)
Quantity 100kU
Source Aspergillus niger
Immunogen Catalase
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C.
Regulatory Status RUO
Recombinant Native
Form Lyophilized