missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Cartridges for RiOs™ Reverse Osmosis Systems

Catalog No. CDRC60201
Change view
Click to view available options
For Use With (Equipment):
Elix™ 10, RiOs™ 16
Elix™ 5, RiOs™ 8
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
CDRC60201 Elix™ 5, RiOs™ 8
CDRC60202 Elix™ 10, RiOs™ 16
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CDRC60201 Supplier MilliporeSigma™ Supplier No. CDRC60201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cartridges for RiOs™ and Elix

Reverse Osmosis Membrane

Specifications

Purification Method Reverse osmosis
Filter Size 2 in.
For Use With (Equipment) Elix™ 5, RiOs™ 8
Model RO
Type Cartridge
Diameter (English) 2 in.