missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Cartridges and Kits for Milli-Q™ Bowl-Style Water Purification Systems

Catalog No. p-169343
Change view
Click to view available options
Type:
Milli-Q™ Cartridge Kit
Milli-Q™ Organicfree Ultrapure Cartridge Kit
Milli-Q™ UF Cartridge Kit
Purification Method:
Ion Exchange, Activated Carbon, Depth Filtration
Ultrafiltration
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Purification Method
CF0F01205 Milli-Q™ Organicfree Ultrapure Cartridge Kit Ion Exchange, Activated Carbon, Depth Filtration
CFIF01205 Milli-Q™ Cartridge Kit Ion Exchange, Activated Carbon, Depth Filtration
CFUF01204 Milli-Q™ UF Cartridge Kit Ultrafiltration
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CF0F01205 Supplier MilliporeSigma™ Supplier No. CF0F01205
Only null left

Pretreatment and purification replacement cartridges

For Milli-Q™ Bowl systems

Specifications

Purification Method Ion Exchange, Activated Carbon, Depth Filtration
For Use With (Equipment) Milli-Q™ bowl-style water purification system
Includes One Super-C™ organics removal cartridge, two ion exchange deionization cartridges, one Organex-Q trace organic removal cartridge, one Millipak™ filter unit
Model Milli-Q™
Type Milli-Q™ Organicfree Ultrapure Cartridge Kit