missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Cartridges and Kits for Milli-Q™ Bowl-Style Water Purification Systems
Description
Pretreatment and purification replacement cartridges
For Milli-Q™ Bowl systems
Specifications
Specifications
| Purification Method | Ion Exchange, Activated Carbon, Depth Filtration |
| For Use With (Equipment) | Milli-Q™ bowl-style water purification system |
| Includes | One Milligard prefiltration cartridge, one Super-C™ organic removal cartridge, two ion exchange deionization cartridges, one Millipak™ filter unit |
| Model | Milli-Q™ |
| Type | Milli-Q™ Cartridge Kit |