Learn More
MilliporeSigma™ Carrez Clarification Reagant Kit
reagent kit for sample preparation in food analysis, 5-fold concentrate
Supplier:  MilliporeSigma™ 1.10537.0001
Catalog No. M1105370001
Only null left
Description
Description
Contains solution I (15g/100mL potassium hexacyanoferrate(II) trihydrate) and solution II (30g/100mL zinc sulfate heptahydrate)
Specifications
Specifications
Carrez Clarification Reagant Kit
Product Suggestions
Product Suggestions