missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem β-Secretase Activity Assay Kit, Fluorogenic

Catalog No. 5657851KIT
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5657851KIT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5657851KIT Supplier MilliporeSigma™ Supplier No. 5657851KIT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Measures beta-secretase (BACE) activity in cell lysates, tissue extracts, or purified enzyme preparations

Specifications

Content And Storage -20°C
Description β-Secretase Activity Assay Kit
Detection Method Fluorogenic
For Use With (Application) Measures beta-secretase (BACE) activity in cell lysates, tissue extracts, or purified enzyme preparations
Includes Extraction Buffer, Reaction Buffer, β-Secretase Substrate, β-Secretase Protein (Positive Control), β-Secretase Inhibitor, and a user protocol.
Quantity 1 Kit