missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Recombinant Mouse Insulin-Like Growth Factor-II, CF

Catalog No. 40725650UG
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40725650UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40725650UG Supplier MilliporeSigma™ Supplier No. 40725650UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recombinant, mouse insulin-like growth factor II (IGF-II) consisting of amino acids 25-91 and expressed in E.Coli

Specifications

Content And Storage -20°C
Formulation Lyophilized from 0.2μm sterile-filtered Acetonitrile, TFA.
Quantity 50 μg
Source E. coli