Learn More
MilliporeSigma™ Calbiochem™ Glutathione Assay Kit II
Measures glutathione from plasma, serum, erythrocyte lysate, tissue samples and cultured cells
Supplier:  MilliporeSigma™ 3541031KIT
Catalog No. 3541031KIT
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
2°C to 8°C
Colorimetric
2X MES Buffer, GSSG Standard, Cofactor Mixture, Enzyme Mixture, DTNB, 96-Well Plate, Plate Cover, and a user protocol. Assay requires Metaphosphoric Acid and Triethanolamine to be supplied by the user.
Glutathione Assay Kit II
Measures glutathione from plasma, serum, erythrocyte lysate, tissue samples and cultured cells
1 Kit (96 Tests)
Product Suggestions
Product Suggestions