missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Caspase-3 Cellular Activity Assay Kit

Catalog No. 2354191KIT
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2354191KIT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2354191KIT Supplier MilliporeSigma™ Supplier No. 2354191KIT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Measures caspase-3 and caspase-3-like activities in cell extract

Specifications

Content And Storage ≤-70°C
Description Caspase-3 Cellular Activity Assay Kit
Detection Method Colorimetric, Fluorometric
Includes Human Recombinant Caspase-3, Cell Lysis Buffer, a DEVD-pNA Colorimetric Substrate, Ac-DEVD-AMC Fluorometric Substrate, Calibration Standard (p-Nitroaniline), AMC Calibration Standard, an Inhibitor, Assay Buffer, one half-volume 96-Well Plate and a user protocol.
Quantity 1 Kit