missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ β amyloid 11-40, Rat and Mouse Recombinant Protein
Specifications
Specifications
| Gene ID (Entrez) | NM_000484.2, NM_201413.1, NM_201414.1 |
| Content And Storage | Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C to -70°C for up to 2 weeks in undiluted aliquots. Avoid freeze/thaw cycles to avoid aggregation. |
| Format | Lyophilized |
| Formulation | White lyophilized powder. Resuspend in 1% NH4OH at a concentration of 1mg/mL. Sonicate for 30 seconds to 1 minute after it has gone into solution. To bring into your solution: After resuspension, add 5x or 10x buffer stock (PBS, TBS) and water to bring to 1X buffer. |
| Gene Symbol | APP, PN-II, AD1, ABETA, CTFgamma, APPI, ABPP, A4, CVAP, PN2, PreA4, AAA |
| Product Type | β amyloid 11-40, rat and mouse |
| Quantity | 500 μg |
| Regulatory Status | RUO - research use only |
| Source | Recombinant |