missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ β amyloid 1-42, Rat and Mouse Recombinant Protein

Catalog No. AG542MI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AG542MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AG542MI Supplier MilliporeSigma™ Supplier No. AG542

Specifications

Gene ID (Entrez) NM_000484.2, NM_201413.1, NM_201414.1
Content And Storage Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C to -70°C for up to 2 weeks in undiluted aliquots. Avoid freeze/thaw cycles to avoid aggregation.
Format Lyophilized
Formulation White lyophilized powder. Resuspend in 1% NH4OH at a concentration of 1mg/mL. Sonicate for 30 seconds to 1 minute after it has gone into solution. To bring into your solution: After resuspension, add 5x or 10x buffer stock (PBS, TBS) and water to bring to 1X buffer.
Gene Symbol APP, PN-II, AD1, ABETA, CTFgamma, APPI, ABPP, A4, CVAP, PN2, PreA4, AAA
Product Type β amyloid 1-42, rat and mouse
Quantity 500 μg
Regulatory Status RUO - research use only
Source Recombinant