Learn More
MilliporeSigma™ Aluminum Oxide 60 TLC Plates
For thin layer chromatography applications.
Supplier: MilliporeSigma™ 1057130001
Description
Aluminiumoxide plates are used to analyze basic and neutral compounds at different pH levels. Depending on the pH range in question, professionals can choose between two kinds of aluminum oxide plates. Under aqueous conditions, basic aluminum oxide plates are best for separating basic compounds, while neutral plates are ideal for separating neutral compounds. Available with or without a fluorescence indicator, our TLC aluminum plates handle a wide variety of applications thanks to neutral and basic aluminum oxides with 60 Å and 150 Å pore sizes.Specifications
| Glass | |
| 60 Å | |
| Aluminum Oxide TLC Plate | |
| Used to Analyze Basic and Neutral Compounds at Different pH Levels | |
| 250 μm | |
| 20 cm |
| Glass | |
| 25/Pk. | |
| 254 nm | |
| Aluminum Oxide 60 F254 Basic | |
| 20 x 20 cm | |
| 20 cm |