missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Hazardous Waste Filtration System
Description
If your waste samples contain non-volatile or semi-volatile substances or metals, use this Hazardous Waste Filtration System for saparation, extratction, and filtration. Developed in collaboration with the US Environmental Protection Agency (EPA), the system is designed specifically for the US EPA Extraction Procedure (EP) Toxicity Test and Toxicity Characteristic Leaching Procedure (TCLP). The device separates solid and liquid phases of waste samples for analysis. Features & Benefits:•Contains interior holder with PTFE coating to prevent heavy metal contamination •Allows easy introduction of bulky samples through wide entry port and removable top plate•Disassembles for easy cleaningApplications:US EPA EP Toxicity Test and Toxicity Characteristic Leaching ProcedureYT31042HW Hazardous Waste Filtration System, 142 mmReplacement Parts:YY2214257 Hand KnobYY2004055 1-1/2 in. TC Silicone Gasket YY2004045 Clamp 1-1/2 in. TCYY2214265 O-ring (2-251), siliconeYY2214253 O-ring (2-251), PTFEYT3014257 Underdrain Support, PTFE-coatedYY2214251 Legs with Caps, 3/16 in. wrenchYY2214252 Handwheel Wrench for 142 mm HolderAccessories:YY2004057 Gasket, 1-1/2 in., PTFE-coated fluoroelastomerXX6700035 Tubing for Pressure Use, 1/2 in. I.D.Membrane Filters:AP4014250 Glass Fiber filter without Binder, AP40, 142 mmAPFF14250 Glass Fiber Filter without Binder, APFF, 142 mmYT310RAHW Rotary Agitator
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Material | Stainless Steel (316), Aluminum, Polypropylene, PTFE |
| Sterility | Non-sterile |
| Type | Hazardous Waste System |
| Height (English) | 16.6 in. |
| Height (Metric) | 42.2 cm |
| Diameter (English) | 7.25 in. |
| Diameter (English) Inside | 7.25 in. |
| Diameter (Metric) Inside | 18.4 cm |