Learn More
MilliporeSigma™ EmbryoMax™ PMEF, Strain CF1, Irradiated, Passage 3
For stem cell culture applications
Supplier:  MilliporeSigma™ PMEFCFX
Catalog No. PMEFCFX
Only null left
Explore available promotions
Description
Description
  • Fibroblasts can be directly plated onto gelatin coated culture vessels
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
-80°C
Primary MEF
Stem cell culture
5 vials
Product Suggestions
Product Suggestions