Learn More
MilliporeSigma™ EmbryoMax™ Embryonic Stem Cell Line, Strain 129/SVEV, Passage 11
For high targeting efficiency and germ line transmission
Supplier:  MilliporeSigma™ CMTI1
Catalog No. CMTI1
Only null left
Description
Description
Specifications
Specifications
Place vials in the vapor phase of liquid nitrogen storage immediately upon receipt until it is convenient to proceed to subculture.
Embryonic Stem Cells (Strain 129/SVEV)
Stem cell culture
2 vials (2.5 x 10^6/Ea.)
Product Suggestions
Product Suggestions