missing translation for 'onlineSavingsMsg'
Learn More

Embelin

Catalog No. AAJ64074MA
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64074MA 10 mg
AAJ64074MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64074MA Supplier Thermo Scientific Chemicals Supplier No. J64074MA

CAS: 550-24-3 | C17H26O4 | 294.39 g/mol

An inhibitor of XIAP (X-linked inhibitor-of-apoptosis protein) that contains antitumor and anti-inflammatory activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • An inhibitor of XIAP (X-linked inhibitor-of-apoptosis protein) that contains antitumor and anti-inflammatory activity

Chemical Identifiers

CAS 550-24-3
Molecular Formula C17H26O4
Molecular Weight (g/mol) 294.39
MDL Number MFCD00016369
InChI Key IRSFLDGTOHBADP-UHFFFAOYSA-N
Synonym embelin, embelic acid, emberine, 2,5-dihydroxy-3-undecyl-1,4-benzoquinone, 2,5-dihydroxy-3-undecyl-p-benzoquinone, 2,5-dihydroxy-3-undecyl-2,5-cyclohexadiene-1,4-dione, unii-shc6u8f5er, embelin, embelia ribes, xiap inhibitor, embelin, shc6u8f5er
PubChem CID 3218
ChEBI CHEBI:4778
IUPAC Name 2,5-dihydroxy-3-undecylcyclohexa-2,5-diene-1,4-dione
SMILES CCCCCCCCCCCC1=C(O)C(=O)C=C(O)C1=O

Specifications

Color Orange
Quantity 10 mg
Sensitivity Light sensitive
Merck Index 14,3555
Formula Weight 294.4
Physical Form Solid
Chemical Name or Material Embelin
EINECSNumber 208-979-8
RTECSNumber DK4230000
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only