Electron Microscopy Sciences Embedding Capsule Holder
Catalog No. 5025729
Fabricated form polypropylene, a direct substitution for the BEEM capsule holders. 15 positions. Fits embedding capsule size 00' (8.5 mm diameter). Sturdy, long lasting, and stackabe.
Catalog No.
5025729
Supplier
Electron Microscopy SciencesSupplier No.
7002201
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Fabricated form polypropylene, a direct substitution for the BEEM capsule holders. 15 positions. Fits embedding capsule size 00' (8.5 mm diameter). Sturdy, long lasting, and stackabe.