missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Elix™ Essential 3, 5, 10, 15 Water Purification System

Catalog No. ZLXEV030US
Change view
Click to view available options
Flow Rate:
10 L/hr.
15 L/hr.
3 L/hr.
5 L/hr.
Includes:
254nm UV lamp
Without UV lamp
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate Includes
ZLXEV030US 3 L/hr. 254nm UV lamp
ZLXEV050US 5 L/hr. 254nm UV lamp
ZLXE0030US 3 L/hr. Without UV lamp
ZLXE0050US 5 L/hr. Without UV lamp
ZLXEV100US 10 L/hr. 254nm UV lamp
ZLXEV150US 15 L/hr. 254nm UV lamp
ZLXE0100US 10 L/hr. Without UV lamp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. ZLXEV030US Supplier MilliporeSigma™ Supplier No. ZLXEV030US

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Choose a Millipore Elix Essential Water Purification System for your lab to ensure consistently-pure, reliable water quality for optimal results.

Ideal for laboratories needing an easy-to-use economical solution that provides a constant and reliable source of Type 2 pure water.

  • Patented Elix™ module, which uses electricity (instead of harsh chemicals and water) to continuously regenerate the resins and remove ions from tap water
  • Intuitive system controls, with system alert and alarm icons shown on a color-coded backlit LCD display
  • High-precision monitoring equipment to ensure precise and reliable measurement of key water quality parameters
  • RFID technology for traceability of Progard™ pretreatment packs
  • New ergonomic pack locking system to make Progard™ pack changes a snap
  • Small system footprint, allowing multiple installation possibilities
  • Has resistivity values greater than 5 Mohm.cm at 25°C and less than 30ppb TOC, exceeding the quality of double distilled water

Feed to laboratory equipment, Preparation of microbiological media, buffer and pH solutions, Histology, Chemical reactions run in water, Manual glassware rinsing

Specifications

Bacterial Content <10 CFU/mL
Feedwater Source Tap
Flow Rate 3 L/hr.
Product Water Type Type 2
Resistivity >5 Mohm.cm
Total Organic Carbon 30 ppb
Display Color Graphic Display
Feedwater Pressure 87 psi (6 bar)
For Use With (Application) Feed to laboratory equipment, preparation of microbiological media, buffer and pH solutions, histology, chemical reactions run in water, manual Glassware rinsing
Includes 254nm UV lamp
Width (English) 10.55 in.
Width (Metric) 26.8 cm
Height (English) 18.5 in.
Height (Metric) 47 cm
Depth (English) 13.35 in.
Depth (Metric) 33.9 cm
Weight (English) 32.95 to 37.51 lb.
Weight (Metric) 12.3 to 14 kg
Electrical Requirements 100/230 V 50/60 Hz
Voltage 100/230 V
Frequency 50/60 Hz
Product Type Elix Essential Water Purification System
Show More Show Less