missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Elix™ Essential 3, 5, 10, 15 Water Purification System

Choose a Millipore Elix Essential Water Purification System for your lab to ensure consistently-pure, reliable water quality for optimal results.

Supplier:  MilliporeSigma™ ZLXE0030US

 View more versions of this product

Catalog No. ZLXE0030US


Only null left

Description

Description

Ideal for laboratories needing an easy-to-use economical solution that provides a constant and reliable source of Type 2 pure water.

  • Patented Elix™ module, which uses electricity (instead of harsh chemicals and water) to continuously regenerate the resins and remove ions from tap water
  • Intuitive system controls, with system alert and alarm icons shown on a color-coded backlit LCD display
  • High-precision monitoring equipment to ensure precise and reliable measurement of key water quality parameters
  • RFID technology for traceability of Progard™ pretreatment packs
  • New ergonomic pack locking system to make Progard™ pack changes a snap
  • Small system footprint, allowing multiple installation possibilities
  • Has resistivity values greater than 5 Mohm.cm at 25°C and less than 30ppb TOC, exceeding the quality of double distilled water

Feed to laboratory equipment, Preparation of microbiological media, buffer and pH solutions, Histology, Chemical reactions run in water, Manual glassware rinsing

Specifications

Chemical Identifiers

<10 CFU/mL
Type 2
30 ppb
87 psi (6 bar)
10.55 in.
18.5 in.
13.35 in.
32.95 to 37.51 lb.
100/230 V 50/60 Hz
50/60 Hz
Tap
>5 Mohm.cm
Color Graphic Display
Feed to laboratory equipment, preparation of microbiological media, buffer and pH solutions, histology, chemical reactions run in water, manual Glassware rinsing
26.8 cm
47 cm
33.9 cm
12.3 to 14 kg
100/230 V
Elix Essential Water Purification System

Specifications

<10 CFU/mL
3 L/hr.
>5 Mohm.cm
Color Graphic Display
Feed to laboratory equipment, preparation of microbiological media, buffer and pH solutions, histology, chemical reactions run in water, manual Glassware rinsing
10.55 in.
18.5 in.
13.35 in.
32.95 to 37.51 lb.
100/230 V 50/60 Hz
50/60 Hz
Tap
Type 2
30 ppb
87 psi (6 bar)
Without UV lamp
26.8 cm
47 cm
33.9 cm
12.3 to 14 kg
100/230 V
Elix Essential Water Purification System
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions