Learn More
Biomatik Corporation Human Serum Amyloid A (SAA) ELISA Kit, 96 tests
Catalog No. 501504495
Encompass
Human Serum Amyloid A (SAA) ELISA Kit for detecting SAA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.66ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P0DJI8. Gene ID: 6288. Target Synonyms: SA-A; SAA1; PIG4; TP53I4; Amyloid protein A; Amyloid fibril protein AA; Serum amyloid A-1 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 501504495 Supplier Biomatik Corporation Supplier No. EKU10220
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Human Serum Amyloid A (SAA) ELISA Kit for detecting SAA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.66ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P0DJI8. Gene ID: 6288. Target Synonyms: SA-A; SAA1; PIG4; TP53I4; Amyloid protein A; Amyloid fibril protein AA; Serum amyloid A-1 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.