Learn More
Biomatik Corporation Mouse Prostaglandin D2 Synthase, Hematopoietic (PTGDS) ELISA Kit, 96 tests
Catalog No. 501503622
Encompass
Mouse Prostaglandin D2 Synthase (PTGDS) ELISA Kit for detecting PTGDS in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JHF7. Gene ID: 54486. Target Synonyms: HPGDS; GSTS; H-PGDS; PDS; PGD2; PGDS; PGDS2; Glutathione S-Transferase Sigma; Glutathione-requiring prostaglandin D synthase; Prostaglandin D2 Synthase, Hematopoietic. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 501503622 Supplier Biomatik Corporation Supplier No. EKU06855
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Mouse Prostaglandin D2 Synthase (PTGDS) ELISA Kit for detecting PTGDS in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JHF7. Gene ID: 54486. Target Synonyms: HPGDS; GSTS; H-PGDS; PDS; PGD2; PGDS; PGDS2; Glutathione S-Transferase Sigma; Glutathione-requiring prostaglandin D synthase; Prostaglandin D2 Synthase, Hematopoietic. Restrictions: For Research Use Only. Not for use in diagnostic procedures.