Learn More
Biomatik Corporation Phospho-Calnexin (Ser583) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291290
Encompass
Phospho-Calnexin (Ser583) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Calnexin (Ser583) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P27824. Gene ID: 821. Target Synonyms: CALX; CANX; IP90; Major histocompatibility complex class I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 502291290 Supplier Biomatik Corporation Supplier No. EKA51092
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Phospho-Calnexin (Ser583) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Calnexin (Ser583) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P27824. Gene ID: 821. Target Synonyms: CALX; CANX; IP90; Major histocompatibility complex class I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.